Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-1,2-phenylenediamine 97.0+%, TCI America™
SDP

Catalog No. D0103100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0103100G 100 g
D010325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0103100G Supplier TCI America Supplier No. D0103100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99-56-9
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007724
InChI Key RAUWPNXIALNKQM-UHFFFAOYSA-N
Synonym 4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
PubChem CID 5111791
ChEBI CHEBI:67116
IUPAC Name 4-nitrobenzene-1,2-diamine
SMILES NC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

Melting Point 201°C
Color Red-Yellow
Quantity 100 g
Formula Weight 153.14
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-1,2-phenylenediamine
EINECSNumber (3)-0402
RTECSNumber ST2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.