Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-1-naphthylamine 98.0+%, TCI America™
SDP

Catalog No. N021325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N021325G 25 g
N02135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N021325G Supplier TCI America Supplier No. N021325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 776-34-1
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00004026
InChI Key BVPJPRYNQHAOPQ-UHFFFAOYSA-N
Synonym 4-nitro-1-naphthylamine, 1-amino-4-nitronaphthalene, 1-naphthalenamine, 4-nitro, 1,4-nitronaphthylamine, 4-nitro-1-naphthamine, 1-naphthylamine, 4-nitro, 4-nitro-1-naphthalenamine, 4-nitro-naphthalen-1-ylamine, 4-nitro-alpha-naphthylamine, 4-nitronaphthylamine
PubChem CID 13057
IUPAC Name 4-nitronaphthalen-1-amine
SMILES C1=CC=C2C(=C1)C(=CC=C2[N+](=O)[O-])N

Specifications

Melting Point 195°C
Quantity 25 g
Formula Weight 188.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-1-naphthylamine
EINECSNumber (4)-0327
RTECSNumber QM4370000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.