Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-2-(trifluoromethyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. N106825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106825G 25 g
N10685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N106825G Supplier TCI America Supplier No. N106825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 320-47-8
Molecular Formula C8H3F3N2O2
Molecular Weight (g/mol) 216.119
MDL Number MFCD01075729
InChI Key AGKQJEFSEQHGTA-UHFFFAOYSA-N
Synonym 4-nitro-2-trifluoromethyl benzonitrile, 2-cyano-5-nitrobenzotrifluoride, 2-trifluoromethyl-4-nitrobenzonitrile, benzonitrile, 4-nitro-2-trifluoromethyl, 4-nitro-2-trifluoromethyl benzenecarbonitrile, pubchem4821, acmc-1ctuy, buttpark 91\04-59, 4-cyano-3-trifluoromethyl nitrobenzene
PubChem CID 2775783
IUPAC Name 4-nitro-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)C#N

Specifications

Melting Point 51°C
Quantity 25 g
UN Number 3439
Formula Weight 216.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-2-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.