Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzamide 98.0+%, TCI America™
SDP

Catalog No. N032325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N032325G 25 g
N0323500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N032325G Supplier TCI America Supplier No. N032325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 619-80-7
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.136
MDL Number MFCD00007994
InChI Key ZESWUEBPRPGMTP-UHFFFAOYSA-N
Synonym p-nitrobenzamide, benzamide, 4-nitro, benzamide, p-nitro, 4-nitro-benzamide, 4-nitro benzamide, 4-carbamoylnitrobenzene, pubchem13419, acmc-209myf, 4-nitrobenzamide
PubChem CID 12091
IUPAC Name 4-nitrobenzamide
SMILES C1=CC(=CC=C1C(=O)N)[N+](=O)[O-]

Specifications

Melting Point 203°C
Color Yellow
Quantity 25 g
Formula Weight 166.14
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzamide
EINECSNumber (3)-1508
RTECSNumber CV5601000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.