Learn More
4-Nitrobenzyl bromide, 97+%
Description
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Nitrobenzyl bromide is used in the synthesis of di and tri-substituted azoles. It is also used in the preparation of N6-Benzyladenosine-5’-uronamides as selective A3 adenosine agonists.
Solubility
It hydrolyzes in water. Soluble in alcohol and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, amines, bases and alcohols.
Specifications
Specifications
| Melting Point | 97°C to 101°C |
| Quantity | 100 g |
| UN Number | UN3261 |
| Beilstein | 742796 |
| Solubility Information | It hydrolyzes in water. Soluble in alcohol and ether. |
| Formula Weight | 216.04 |
| Percent Purity | ≥97% |
| Chemical Name or Material | 4-Nitrobenzyl bromide |
Safety and Handling
| Hazard Category | H314-H318 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 202-820-6 |
| RTECSNumber | XS7967000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.