Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. p-7133806 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04355G 5 g
N043525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04355G Supplier TCI America Supplier No. N04355G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18600-42-5
Molecular Formula C7H9ClN2O2
Molecular Weight (g/mol) 188.611
MDL Number MFCD00012863
InChI Key SMIXZZMSWYOQPW-UHFFFAOYSA-N
Synonym 4-nitrobenzylamine hydrochloride, 4-nitrophenyl methanamine hydrochloride, p-nitrobenzylamine hydrochloride, 4-nitrobenzylamine hcl, pubchem12455, acmc-209enh, 4-nitrobenzylamine hcl salt, ksc493k2b, p-nitrobenzylaminehydrochloride, 4-nitrobenzylaminehydrochloride
PubChem CID 11252467
IUPAC Name (4-nitrophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC=C1CN)[N+](=O)[O-].Cl

Specifications

Melting Point 265°C
Color White-Yellow
Quantity 5 g
Formula Weight 188.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzylamine Hydrochloride
RTECSNumber DP6705000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.