Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitroindole 98.0+%, TCI America™
SDP

Catalog No. N06281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06281G 1 g
N06285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06281G Supplier TCI America Supplier No. N06281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4769-97-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00010056
InChI Key LAVZKLJDKGRZJG-UHFFFAOYSA-N
Synonym 4-nitroindole, indole, 4-nitro, 1h-indole, 4-nitro, pubchem7440, 4-nitro-1 h-indole, #, maybridge1_006408, acmc-209ka5, ksc236o2l
PubChem CID 145766
IUPAC Name 4-nitro-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)[N+](=O)[O-]

Specifications

Melting Point 206°C
Quantity 1 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitroindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.