Learn More
Cayman Chemical RubIadIn-1 methyl ether 10mg
Supplier: Cayman Chemical 3494010 MG
An anthraquinone with diverse biological activities; generates superoxide radicals in isolated human neutrophils and monocytes at 20 UG/ml, an effect that can be enhanced by UV radiation; inhibits M-CSF- and RANKL-induced differentiation of osteoclasts at 0.1, 1, and 10 µM; inhibits C. tropicalis biofilm formation (MIC = 31.3 UG/ml for a clinical isolate); reduces LPS-induced increases in BALF levels of TNF-α, IL-6, and IFN-γ in a mouse model of acute lung injury at 30 mg/kg
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.