Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl Isocyanate (contains varying amounts of polymers) 98.0+%, TCI America™
SDP

Catalog No. I012925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I012925G 25 g
I01295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I012925G Supplier TCI America Supplier No. I012925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100-28-7
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
MDL Number MFCD00007306
InChI Key GFNKTLQTQSALEJ-UHFFFAOYSA-N
Synonym 4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
PubChem CID 66012
IUPAC Name 1-isocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=O)[N+](=O)[O-]

Specifications

Melting Point 58°C
Color Yellow
Boiling Point 162°C
Quantity 25 g
Formula Weight 164.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl Isocyanate (contains varying amounts of polymers)
RTECSNumber DA3692500
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.