Learn More
Sigma Organic Chemistry 4-Nitrophenylacetic aci | 100G | 104-03-0 | MFCD00007383
Supplier: Sigma Organic Chemistry N20204100G
4-Nitrophenylacetic aci , 100G
About this item:
CAS #: 104-03-0
MDL #: MFCD00007383
Purity: 99%
Chemical Formula: C8H7NO4
Molecular Weight: 181.15
UNSPSC Code: 12352100
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.