Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenylhydrazine mono and dihydrochloride, 98%

Catalog No. AAL0927304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0927304 2 g
AAL0927309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0927304 Supplier Thermo Scientific Chemicals Supplier No. L0927304
Add to Cart

C6H7N3O2·xHCl

4-Nitrophenylhydrazine mono and dihydrochloride exhibits potential anticancer activity. It acts as a derivatizing agent for carbonyl compounds. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenylhydrazine mono and dihydrochloride exhibits potential anticancer activity. It acts as a derivatizing agent for carbonyl compounds. It is also used as pharmaceutical intermediates.

Solubility
Soluble in water, DMSO and methanol.

Notes
Hygroscopic. Incompatible materials are Strong oxidizing agents and moisture. Store under dry inert gas. Protect from humidity nd water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6H7N3O2·xHCl
MDL Number MFCD00040592

Specifications

Melting Point ∽200°C (decomposition)
Quantity 2 g
UN Number UN2811
Sensitivity Hygroscopic
Solubility Information Soluble in water,DMSO and methanol.
Formula Weight 153.14(f.b.)
Percent Purity 98%
Chemical Name or Material 4-Nitrophenylhydrazine mono and dihydrochloride
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.