Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophthalic Anhydride 98.0+%, TCI America™
SDP

Catalog No. N0246100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0246100G 100 g
N024625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0246100G Supplier TCI America Supplier No. N0246100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5466-84-2
Molecular Formula C8H3NO5
Molecular Weight (g/mol) 193.114
MDL Number MFCD00005922
InChI Key MMVIDXVHQANYAE-UHFFFAOYSA-N
Synonym 4-nitrophthalic anhydride, 5-nitroisobenzofuran-1,3-dione, phthalic anhydride, 4-nitro, 1,3-isobenzofurandione, 5-nitro, 4-nitrophthalic acid anhydride, 5-nitrophthalic anhydride, unii-sqn32ua63n, ccris 4684, 5-nitro-1,3-isobenzofurandione, sqn32ua63n
PubChem CID 230976
IUPAC Name 5-nitro-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)OC2=O

Specifications

Melting Point 120°C
Color Yellow
Boiling Point 197°C
Quantity 100 g
Formula Weight 193.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophthalic Anhydride
RTECSNumber TI3328000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.