Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophthalimide 98.0+%, TCI America™
SDP

Catalog No. N024725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N024725G 25 g
N0247500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N024725G Supplier TCI America Supplier No. N024725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89-40-7
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00005884
InChI Key ANYWGXDASKQYAD-UHFFFAOYSA-N
Synonym 4-nitrophthalimide, 5-nitroisoindoline-1,3-dione, phthalimide, 4-nitro, 5-nitro-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-nitro, 5-nitrophthalimide, unii-26na19ui3u, 5-nitro-2,3-dihydro-1h-isoindole-1,3-dione, ccris 4685, 5-nitro-2h-benzo c azoline-1,3-dione
PubChem CID 6969
IUPAC Name 5-nitroisoindole-1,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)NC2=O

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 25 g
Formula Weight 192.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophthalimide
EINECSNumber (5)-0081
RTECSNumber TI5625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.