Learn More
Cayman Chemical 4-O-MethylhonkIol 1mg
SDP
Supplier:  Cayman Chemical 110891 MG
Encompass_Preferred
CAY10502 s a potent nhbtor of cPLA2a wth an C50 value of 4 3 nM for the purfed enzyme from human platelets 14126 t nhbts arachdonc acd moblzaton from A23187-stmulated or TPA-stmulated human platelets wth C50 values of 570 and 0 9 nM respectvely 14126
Catalog No. 50-295-2062
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.