Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 4-Octyl Itaconate
GREENER_CHOICE

Catalog No. p-7161590
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-621-0 10 mg
66-625-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 66-621-0 Supplier Tocris Bioscience™ Supplier No. 6662/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nrf2 activator; cell permeable

4-Octyl itaconate activates Nrf2. Inhibits LPS-induced increases in IL-1β mRNA, HIF-1α and IL-10 in macrophages. Decreases cytokine production in response to LPS in mice and prolongs survival. Cell permeable.

Specifications

Chemical Name or Material 2-Methylenebutanedioic acid 4-octyl ester
CAS 3133-16-2
Quantity 10 mg
Molecular Formula C13H22O4
Formula Weight 242.31
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.