Learn More
STEMCELL Technologies 4-Octyl Itaconate, Size: 50 mg
SDP
Supplier:  STEMCELL Technologies 74192
Encompass_Preferred
4-Octyl Itaconate is a cell-permeable derivative of itaconate, a metabolite produced during the Krebs cycle. It is a potent activator of nuclear factor erythroid 2-related factor 2 (Nrf2) via alkylation of the inhibitor of Nrf2 (Keap1), thereby promoting Nrf2-mediated transcriptional activation and downstream anti-oxidant and anti-inflammatory properties (Mills et al.; Tang et al.).IMMUNOLOGY· The itaconate pathway (IRG1-itaconate-SDH) plays a pivotal role in up-regulating tolerance and inhibiting the induction of trained immunity (Domínguez-Andrés et al.).· Counteracts the proinflammatory signals of succinate by inhibiting succinate dehydrogenase (SDH) and activating Nrf2 to limit inflammation in mouse and human macrophages (Lampropoulou et al.; Mills et al.).· Activates Nrf2 signaling in human macrophages, which has been shown to inhibit the production of pro-inflammatory cytokines including TNF-α, IL-1β, and IL-6 (Tang et al.).
Catalog No. 50-197-6196
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.