Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Octylphenol, MilliporeSigma™ Supelco™

Catalog No. 111013374 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3374 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3374 Supplier MilliporeSigma Supelco Supplier No. 442850

4-Octylphenol belongs to the class of alkylphenolpolyethoxylates, commonly used as non-ionic surfactants in a variety of commercial and industrial applications.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 1806-26-4
Density 0.961 g/mL (at 25°C)
Flash Point 113°C
Physical Form Neat
Linear Formula CH3(CH2)7C6H4 OH
MDL Number MFCD00036134
Melting Point 44°C to 45°C
Boiling Point 150°C (4 mmHg)
Type Organic Pollutant Standard
Quantity 500 mg
Formula Weight 206.32
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.