Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Oxatetradecanoic acid, 99%, Thermo Scientific™

Catalog No. AAJ67471MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67471MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67471MA Supplier Thermo Scientific Chemicals Supplier No. J67471MA

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7420-16-8
Molecular Formula C13H26O3
Molecular Weight (g/mol) 230.35
MDL Number MFCD00214355
InChI Key ZYLHDTBQHXSLEZ-UHFFFAOYSA-N
Synonym 4-oxatetradecanoic acid, 3-decyloxy propanoic acid, 4-oxymyristic acid, 3-decyloxypropanoic acid, 3-decanoxypropanoic acid, ambscpod_05/0191, propanoic acid,3-decyloxy, 4-om, bml3-c07, propanoic acid, 3-decyloxy
PubChem CID 1754
IUPAC Name 3-(decyloxy)propanoic acid
SMILES CCCCCCCCCCOCCC(O)=O

Specifications

Quantity 10 mg
Formula Weight 230.35
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4-Oxatetradecanoic acid
storageNote1 -30° to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.