Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 4-Oxo-2-nonenal, TRPA1 Activator, >98% Purity, Liquid, 5 mg

Catalog No. 502707178
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7178 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7178 Supplier Abcam Supplier No. AB1208785MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent lipid peroxidation product and TRPA1 activator with >98% purity.

  • Molecular weight: 154.21 g/mol
  • Purity: >98%
  • Form: Liquid
  • Storage buffer: Constituents: 99.5% Methyl acetate, 0.5% (E)-4-oxonon-2-enal
  • CAS number: 103560-62-9
  • Activity: Lipid peroxidation product that modifies histidine and lysine residues on proteins, causes protein cross-linking, and acts as a TRPA1 activator (EC50 = 1.9 μM)
  • Stability: Active in vivo
  • Storage conditions: Store at -80°C
  • Shipping conditions: Dry Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule TRPA1
Shipping Condition Dry Ice
Content And Storage Store at -80°C.
Form Liquid
Format Vial
Formulation 99.5% Methyl acetate, 0.5% (E)-4-oxonon-2-enal
Quantity 5 mg
For Use With (Application) TRPA1 activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.