Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Pentenoyl chloride

Catalog No. 46847925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-847-925ML 25 mL
46-847-95ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-847-925ML Supplier Sigma Aldrich Supplier No. 46847925ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-pentenoyl chloride has been identified as a key impurity in 5-chlorovaleroyl chloride (5-cvc). 4-pentenoyl chloride can be synthesized by reacting thionyl chloride and 4-pentenoic acid.4-pentenoyl chloride may be used in the preparation of:
  • D-pro-L-derived cyclic peptides
  • 4-pentenoylcobalt tricarbonyl
  • N-4-pentenoyl-L-cysteine methyl ester

Specifications

Boiling Point 125°C (lit.)
CAS 39716-58-0
Quantity 25 mL
Molecular Formula C5H7ClO
Linear Formula H2C=CHCH2CH2COCl
MDL Number MFCD00799572
Molecular Weight (g/mol) 118.56
Percent Purity 98%
Density 1.074 g/mL (at 25°C (literature))
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.