Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-phenoxybenzylamine

Catalog No. CDS004637 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS004637 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS004637 Supplier Sigma Aldrich Supplier No. CDS004637100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trans-2-pentenal (pentenal) is an α,α-unsaturated aldehyde that increases [ca2+] in cultured neonatal rat trigeminal ganglion (tg) neurons
  • The gas-phase reactions of no3 radicals with pentenal in pure n2, using a relative rate technique, was studied

Specifications

CAS 107622-80-0
Molecular Formula C13H13NO
Linear Formula C13H13NO
MDL Number MFCD01310836
Quantity 5 g
Molecular Weight (g/mol) 199.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.