Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Phenoxymethyl)phenylboronic acid, 97%, Thermo Scientific™

Catalog No. CC63712CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC63712CB 250 mg Amber Glass Bottle
CC63712DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC63712CB Supplier Thermo Scientific Chemicals Supplier No. CC63712CB
Add to Cart

Chemical Identifiers

CAS 397843-61-7
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.05
MDL Number MFCD09966155
InChI Key IXDHTLPOOZIZJX-UHFFFAOYSA-N
Synonym 4-phenoxymethyl phenylboronic acid, 4-phenoxymethyl phenyl boronic acid, acmc-1aegd, 4-phenoxymethyl phenyl boronicacid
PubChem CID 10955308
IUPAC Name [4-(phenoxymethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(COC2=CC=CC=C2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 228.05
Percent Purity 97%
Chemical Name or Material 4-(Phenoxymethyl)phenylboronic acid
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.