Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Phenyl-1-butene

Catalog No. P2060250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2060250ML 50 mL
P2060210ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2060250ML Supplier Sigma Aldrich Supplier No. P2060250ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Phenyl-1-butene

Specifications

CAS 768-56-9
Density 0.88 g/mL (at 25°C (literature))
Boiling Point 175°C to 177°C (lit.)
Molecular Formula C10H12
Refractive Index n20/D 1.507 (literature)
Linear Formula C6H5CH2CH2CH=CH2
MDL Number MFCD00026111
Quantity 50 mL
Molecular Weight (g/mol) 132.2
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.