Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-2-(4-pyridyl)thiazole-5-carboxamidoxime, 97%, Thermo Scientific™

Catalog No. AAH5839903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5839903 1 g
AAH5839906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5839903 Supplier Thermo Scientific Chemicals Supplier No. H5839903

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1858264-29-5
Molecular Formula C15H12N4OS
Molecular Weight (g/mol) 296.348
MDL Number MFCD22988950
InChI Key VKJURGBMUDKEDZ-BUHFOSPRSA-N
Synonym 4-phenyl-2-4-pyridyl thiazole-5-carboxamidoxime
PubChem CID 121235639
IUPAC Name (E)-nitroso-[4-phenyl-2-(1H-pyridin-4-ylidene)-1,3-thiazol-5-ylidene]methanamine
SMILES C1=CC=C(C=C1)C2=NC(=C3C=CNC=C3)SC2=C(N)N=O

Specifications

Quantity 1 g
Formula Weight 296.34
Percent Purity 97%
Chemical Name or Material 4-Phenyl-2-(4-pyridyl)thiazole-5-carboxamidoxime
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.