Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylethynylphthalic Anhydride 98.0+%, TCI America™
SDP

Catalog No. P143725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P143725G 25 g
P14375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P143725G Supplier TCI America Supplier No. P143725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 119389-05-8
Molecular Formula C16H8O3
Molecular Weight (g/mol) 248.237
MDL Number MFCD01862632
InChI Key UPGRRPUXXWPEMV-UHFFFAOYSA-N
PubChem CID 11053989
IUPAC Name 5-(2-phenylethynyl)-2-benzofuran-1,3-dione
SMILES C1=CC=C(C=C1)C#CC2=CC3=C(C=C2)C(=O)OC3=O

Specifications

Melting Point 152°C
Color Yellow
Quantity 25 g
Formula Weight 248.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylethynylphthalic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.