Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylimidazole 98.0+%, TCI America™
SDP

Catalog No. P087725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P087725G 25 g
P08775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P087725G Supplier TCI America Supplier No. P087725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 670-95-1
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00005197
InChI Key XHLKOHSAWQPOFO-UHFFFAOYSA-N
Synonym 4-phenylimidazole, 4-phenyl-1h-imidazole, 1h-imidazole, 4-phenyl, 4 5-phenylimidazole, imidazole, 4-phenyl, 4-phenyl imidazole, chembl14145, pim, 4-phenyl-1h-imidazol, 5-phenylimidazole
PubChem CID 69590
IUPAC Name 5-phenyl-1H-imidazole
SMILES C1=CC=C(C=C1)C2=CN=CN2

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 25 g
Formula Weight 144.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.