Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-phenyltetrahydropyran-4-yl)methylamine, 95%, Thermo Scientific™

Catalog No. CC69113DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC69113DA 1 g Amber Glass Bottle
CC69113CB 250 mg Amber Glass Bottle
CC69113DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC69113DA Supplier Thermo Scientific Chemicals Supplier No. CC69113DA
Add to Cart

Chemical Identifiers

CAS 14006-32-7
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD02946795
InChI Key IPYDNTFWMDEOBW-UHFFFAOYSA-N
Synonym 4-phenyloxan-4-yl methanamine, 4-phenyltetrahydro-2h-pyran-4-yl methanamine, 4-phenyltetrahydropyran-4-yl methylamine, 1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, c-4-phenyl-tetrahydro-pyran-4-yl-methylamine, 4-phenyltetrahydro-2h-pyran-4-yl methylamine, 1-4-phenyloxan-4-yl methanamine, 2h-pyran-4-methanamine, tetrahydro-4-phenyl, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine, 4-phenyl-tetrahydro-2h-pyran-4-methylamine
PubChem CID 6483779
SMILES NCC1(CCOCC1)C1=CC=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (4-phenyloxan-4-yl)methanamine
Formula Weight 191.27
Percent Purity 95%
Chemical Name or Material (4-phenyltetrahydropyran-4-yl)methylamine
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.