Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylurazole 98.0+%, TCI America™
SDP

Catalog No. P116025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P116025G 25 g
P11605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P116025G Supplier TCI America Supplier No. P116025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15988-11-1
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD00005226
InChI Key GOSUFRDROXZXLN-UHFFFAOYSA-N
Synonym 4-phenylurazole, urazole, 4-phenyl, 1,2,4-triazolidine-3,5-dione, 4-phenyl, bicarbamimide, n-phenyl, 4-phenyl urazole, 4-phenylurazol, 4-phenyl-urazole, acmc-1c52y, ksc297q5n
PubChem CID 85229
IUPAC Name 4-phenyl-1,2,4-triazolidine-3,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)NNC2=O

Specifications

Melting Point 209°C
Color White
Quantity 25 g
Formula Weight 177.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylurazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.