Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4α-Phorbol 12-Myristate 13-Acetate, MP Biomedicals™

Catalog No. MP218388205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218388205 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP218388205 Supplier MP Biomedicals, Inc Supplier No. 0218388205
Add to Cart
Add to Cart

Synonym: Phorbol 12-Myristate 13-Acetate, 12-O-Tetradecanoylphorbol-13-Acetate, Tetradecanoylphorbol Acetate

Negative control for phorbol ester activation of protein kinase C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63597-44-4

Specifications

Quantity 5 mg
Physical Form Solid
Purity ≥99%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.