Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(piperidin-1-ylmethyl)benzaldehyde, 95%, Thermo Scientific™

Catalog No. CC45504DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC45504DA 1 g
CC45504CB 250 mg
CC45504DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC45504DA Supplier Thermo Scientific Chemicals Supplier No. CC45504DA
Add to Cart

Chemical Identifiers

CAS 471929-86-9
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
InChI Key WWBOSCKXPMBVLG-UHFFFAOYSA-N
PubChem CID 7164649
IUPAC Name 4-(piperidin-1-ylmethyl)benzaldehyde
SMILES C1CCN(CC1)CC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 95%
Quantity 1 g
Synonym 4-piperidin-1-ylmethyl benzaldehyde, 4-1-piperidinylmethyl benzaldehyde, benzaldehyde,4-1-piperidinylmethyl, 4-piperidylmethyl benzaldehyde, 4-piperidinomethyl benzaldehyde, benzaldehyde, 4-1-piperidinylmethyl
Formula Weight 203.28
Percent Purity 95%
Chemical Name or Material 4-(Piperidin-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.