Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-piperidineacetic acid hydrochloride

Catalog No. CDS007853 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS007853 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS007853 Supplier Sigma Aldrich Supplier No. CDS007853250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reaction of 1-chloro-2-methyl-2-propanol on oxygen-containing Ag(110) surfaces has been investigated by synchrotron-based temperature-programmed X-ray photoelectron spectroscopy.1-Chloro-2-methyl-2-propanol was used in the synthesis and characterization of new polyfunctional thiols.

Specifications

Molecular Formula C7H14ClNO2
Linear Formula C7H14ClNO2
MDL Number MFCD03414187
Quantity 50 mL
Molecular Weight (g/mol) 179.64
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.