Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Piperidinecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. I0256100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0256100G 100 g
I025625G 25 g
I0256500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0256100G Supplier TCI America Supplier No. I0256100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 498-94-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006004
InChI Key SRJOCJYGOFTFLH-UHFFFAOYSA-N
Synonym isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
PubChem CID 3773
IUPAC Name piperidine-4-carboxylic acid
SMILES OC(=O)C1CCNCC1

Specifications

Melting Point 336°C
Quantity 100 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Piperidinecarboxylic Acid
EINECSNumber (5)-0770
RTECSNumber NS5150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.