Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Piperidinoaniline, 97%, Thermo Scientific™

Catalog No. BTB05830EA
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB05830EA 10 g Amber Glass Bottle
BTB05830DA 1 g Amber Glass Bottle
BTB05830EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB05830EA Supplier Thermo Scientific Chemicals Supplier No. BTB05830EA
Add to Cart

Chemical Identifiers

CAS 2359-60-6
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.263
MDL Number MFCD00051688
InChI Key TVOSOIXYPHKEAR-UHFFFAOYSA-N
Synonym 4-piperidinoaniline, 4-piperidin-1-yl-phenylamine, 4-piperidin-1-yl aniline, 4-1-piperidinyl aniline, 1-4-aminophenyl piperidine, benzenamine, 4-1-piperidinyl, n-4-aminophenyl piperidine, 4-1-piperidyl aniline, 4-1-piperidino aniline, 4-piperidylphenylamine
PubChem CID 413501
IUPAC Name 4-piperidin-1-ylaniline
SMILES C1CCN(CC1)C2=CC=C(C=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 4-Piperidinoaniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.