Learn More
Agilent BioTek 4-position Slide Adapter
GREENER_CHOICE
Catalog No. BT1650516 Shop All Agilent Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Agilent BioTek Microplate Washers and Readers
Catalog No. For Use With (Equipment)
BT1650516 Agilent BioTek Microplate Washers and Readers
1 options
Catalog No. BT1650516 Supplier Agilent BioTek Supplier No. 1650516
Add to Cart
Add to Cart
4-position slide adapter is compatible with BioStack 3 and BioStack 4, to automate handling of up to 100 slides (25 adapters per stack)
  • Adapter can hold up to 4 microscope slides
TRUSTED_SUSTAINABILITY
Specifications
Product Type Slide Adapter
For Use With (Equipment) Agilent BioTek Microplate Washers and Readers
Holds 4 Microscope Slides