Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propionylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. P121225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P121225G 25 g
P12125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P121225G Supplier TCI America Supplier No. P121225G

Chemical Identifiers

CAS 37940-57-1
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00027015
InChI Key JMBGDQSXJHLFTO-UHFFFAOYSA-N
PubChem CID 95230
IUPAC Name 1-(4-phenylphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 96°C
Color Yellow
Boiling Point 344°C
Quantity 25 g
Synonym 4′-Phenylpropiophenone
Formula Weight 210.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Propionylbiphenyl
RTECSNumber UH4400550
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.