Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propylphenol 99.0+%, TCI America™
SDP

Catalog No. P069825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P069825G 25 g
P06985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P069825G Supplier TCI America Supplier No. P069825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 645-56-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002395
InChI Key KLSLBUSXWBJMEC-UHFFFAOYSA-N
Synonym 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
PubChem CID 12580
ChEBI CHEBI:34434
IUPAC Name 4-propylphenol
SMILES CCCC1=CC=C(C=C1)O

Specifications

Color White-Yellow
Boiling Point 232°C
Quantity 25 g
UN Number 2430
Formula Weight 136.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Propylphenol
EINECSNumber (3)-0503&(4)-0057
RTECSNumber SM8610000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.