Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propylpyridine 97.0+%, TCI America™
SDP

Catalog No. P066925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P066925ML 25 mL
P06695ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P066925ML Supplier TCI America Supplier No. P066925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1122-81-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00014639
InChI Key JAWZAONCXMJLFT-UHFFFAOYSA-N
Synonym 4-n-propylpyridine, pyridine, 4-propyl, 4-n-propylpylpyridine, 4-n-propyl-pyridine, 4-propyl-pyridin, 4-propyl pyridine, pubchem15021, 1-4-pyridyl propane, 1-pyridin-4-yl propane, 4-n-propylpyridine 97+%
PubChem CID 70738
IUPAC Name 4-propylpyridine
SMILES CCCC1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 190°C
Quantity 25 mL
Formula Weight 121.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Propylpyridine
EINECSNumber (9)-1843
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.