Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridylamidoxime 98.0+%, TCI America™
SDP

Catalog No. P25911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25911G 1 g
P25915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P25911G Supplier TCI America Supplier No. P25911G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1594-57-6
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.14
MDL Number MFCD00125873
InChI Key ZUUATXHRJFHSGO-UHFFFAOYSA-N
Synonym n'-hydroxypyridine-4-carboximidamide, pyridine-4-amidoxime, 4-pyridylamidoxime, n-hydroxyisonicotinamidine, n-hydroxy-isonicotinamidine, isonicotinamidoxime, isonicotinohydroxamamide, 4-pyridinecarboxamidoxime, n-hydroxyisonicotinimidamide, z-n'-hydroxyisonicotinimidamide
PubChem CID 5355713
IUPAC Name (Z)-N'-hydroxypyridine-4-carboximidamide
SMILES N\C(=N/O)C1=CC=NC=C1

Specifications

Melting Point 205°C
Color White-Yellow
Quantity 1 g
Formula Weight 137.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyridylamidoxime
RTECSNumber NS1050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.