Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Pyrrolidin-1-ylmethyl)aniline, 97%, Thermo Scientific™

Catalog No. CC55614DA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC55614DA 1 g
CC55614CB 250 mg
CC55614EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC55614DA Supplier Thermo Scientific Chemicals Supplier No. CC55614DA
Add to Cart

Chemical Identifiers

CAS 142335-64-6
Molecular Formula C11H17N2
Molecular Weight (g/mol) 177.27
MDL Number MFCD03724739
InChI Key SFEAIUCOZWDYMJ-UHFFFAOYSA-O
Synonym 4-pyrrolidin-1-ylmethyl aniline, 4-pyrrolidin-1-ylmethyl-phenylamine, benzenamine, 4-1-pyrrolidinylmethyl, 4-1-pyrrolidinylmethyl aniline, 4-pyrrolidin-1-ylmethyl-aniline, 4-pyrrolidinylmethyl phenylamine, 1-4-aminobenzyl pyrrolidine, 4-pyrrolizinomethyl aniline, 4-pyrrolidinyl-methyl aniline, cbi-bb zero/006543
PubChem CID 776852
IUPAC Name 4-(pyrrolidin-1-ylmethyl)aniline
SMILES NC1=CC=C(C[NH+]2CCCC2)C=C1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 176.26
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.