Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyrrolidinopyridine 98.0+%, TCI America™
SDP

Catalog No. P093925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P093925G 25 g
P09395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P093925G Supplier TCI America Supplier No. P093925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2456-81-7
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD00006415
InChI Key RGUKYNXWOWSRET-UHFFFAOYSA-N
Synonym 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine
PubChem CID 75567
IUPAC Name 4-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=CC=NC=C2

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 170°C
Quantity 25 g
UN Number 2923
Formula Weight 148.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyrrolidinopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.