Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B331225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B331225G 25 g
B33125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B331225G Supplier TCI America Supplier No. B331225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57699-45-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00040800
InChI Key VWSFZYXXQDKXKQ-UHFFFAOYSA-N
Synonym 4-tert-butoxy benzaldehyde, 4-tert-butoxybenzaldehyde, p-tert-butoxybenzaldehyde, benzaldehyde, 4-1,1-dimethylethoxy, 4-2-methylpropan-2-yl oxy benzaldehyde, 4-t-butoxybenzaldehyde, acmc-209lzx, 4-t-butyloxybenzaldehyde, 4-t-butoxy benzaldehyde, p-tert.-butoxy-benzaldehyde
PubChem CID 93774
IUPAC Name 4-[(2-methylpropan-2-yl)oxy]benzaldehyde
SMILES CC(C)(C)OC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 93°C
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butoxybenzaldehyde
EINECSNumber (9)-1166
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.