Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(tert-Butoxycarbonylamino)methyl]pyridine 98.0+%, TCI America™
SDP

Catalog No. B212625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B212625G 25 g
B21265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B212625G Supplier TCI America Supplier No. B212625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111080-65-0
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.261
MDL Number MFCD01321573
InChI Key KXCFJDZVQLRYSI-UHFFFAOYSA-N
Synonym tert-butyl pyridin-4-ylmethyl carbamate, n-boc-4-aminomethylpyridine, 4-tert-butoxycarbonylamino methyl pyridine, tert-butyl n-pyridin-4-ylmethyl carbamate, pyridin-4-ylmethyl carbamic acid tert-butyl ester, tert-butyl pyridin-4-ylmethylcarbamate, 4-t-butyloxycarbonylaminomethyl-pyridine, 4-tert-butyloxycarbonylaminomethyl pyridin, 4-tert-butyloxycarbonylaminomethyl pyridine
PubChem CID 1512532
IUPAC Name tert-butyl N-(pyridin-4-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=NC=C1

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(tert-Butoxycarbonylamino)methyl]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.