Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(tert-Butoxycarbonylamino)pyridine 98.0+%, TCI America™
SDP

Catalog No. B333125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B333125G 25 g
B33315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B333125G Supplier TCI America Supplier No. B333125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98400-69-2
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.234
MDL Number MFCD02179232
InChI Key DRZYCRFOGWMEES-UHFFFAOYSA-N
Synonym 4-boc-amino pyridine, 4-tert-butoxycarbonylamino pyridine, tert-butyl pyridin-4-ylcarbamate, tert-butyl n-pyridin-4-yl carbamate, tert-butyl n-4-pyridyl carbamate, carbamic acid, 4-pyridinyl-, 1,1-dimethylethyl ester, pyridin-4-yl-carbamic acid tert-butyl ester, pubchem20060, acmc-209sa9, ksc495m3f
PubChem CID 9990210
IUPAC Name tert-butyl N-pyridin-4-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1=CC=NC=C1

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(tert-Butoxycarbonylamino)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.