Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene 94.0+%, TCI America™
SDP

Catalog No. B23101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23101G 1 g
B23105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23101G Supplier TCI America Supplier No. B23101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 210706-03-9
Molecular Formula C56H72O12S4
Molecular Weight (g/mol) 1065.42
MDL Number MFCD06797059
InChI Key IGVIIKFHGZNQFL-UHFFFAOYSA-N
Synonym Tetra-tert-butyl[tetrakis(ethoxycarbonylmethoxy)]tetrathiacalix[4]arene
PubChem CID 5248766
IUPAC Name ethyl 2-{[5,11,17,23-tetra-tert-butyl-26,27,28-tris(2-ethoxy-2-oxoethoxy)-2,8,14,20-tetrathiapentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3(28),4,6,9(27),10,12,15,17,19(26),21,23-dodecaen-25-yl]oxy}acetate
SMILES CCOC(=O)COC1=C2SC3=CC(=CC(SC4=C(OCC(=O)OCC)C(SC5=C(OCC(=O)OCC)C(SC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(=O)OCC)C(C)(C)C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 1065.42
Percent Purity ≥94.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.