Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butyl-o-xylene 98.0+%, TCI America™
SDP

Catalog No. B1338500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1338500G 500 g
B133825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1338500G Supplier TCI America Supplier No. B1338500G

Chemical Identifiers

CAS 7397-06-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00038277
InChI Key QRPPSTNABSMSCS-UHFFFAOYSA-N
Synonym 4-tert-butyl-o-xylene, 4-tert-butyl-1,2-dimethylbenzene, 4-t-butyl-o-xylene, benzene, 4-1,1-dimethylethyl-1,2-dimethyl, acmc-209ot4, o-xylene, 4-tert-butyl, as-p2, 1,2-dimethyl-4-t-butylbenzene, 3,4-dimethyl-tert-butylbenzene
PubChem CID 81881
IUPAC Name 4-tert-butyl-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)C(C)(C)C)C

Specifications

Color Colorless
Boiling Point 211°C
Quantity 500 g
Formula Weight 162.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butyl-o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.