Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylbenzaldehyde 95.0+%, TCI America™
SDP

Catalog No. B1760100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1760100G 100 g
B176025G 25 g
B1760500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1760100G Supplier TCI America Supplier No. B1760100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 939-97-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00035742
InChI Key OTXINXDGSUFPNU-UHFFFAOYSA-N
Synonym 4-tert-butyl benzaldehyde, p-t-butylbenzaldehyde, p-tertbutyl benzaldehyde, benzaldehyde, 4-1,1-dimethylethyl, p-tert-butylbenzaldehyde, 4-t-butylbenzaldehyde, 4-tert-butyl-benzaldehyde, unii-qxf0qy8503, 4-t-butyl benzaldehyde, benzaldehyde, p-tert-butyl
PubChem CID 70324
IUPAC Name 4-tert-butylbenzaldehyde
SMILES CC(C)(C)C1=CC=C(C=O)C=C1

Specifications

Color Yellow
Boiling Point 246°C
Quantity 100 g
UN Number 3082
Formula Weight 162.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzaldehyde
EINECSNumber (3)-2663
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.