Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylbenzeneboronic acid, 97%

Catalog No. p-7027563
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2440803 1 g
AAB2440806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2440803 Supplier Thermo Scientific Chemicals Supplier No. B2440803
Only null left
Add to Cart

CAS: 123324-71-0 | C10H15BO2 | 178.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123324-71-0
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.04
MDL Number MFCD01009697
InChI Key MNJYZNVROSZZQC-UHFFFAOYSA-N
Synonym 4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid, 4-tert-butyl-phenyl boronic acid, 4-tert-butylphenyl boranediol, p-t-butylphenylboronic acid, 4-tertbutylphenylboronic acid
PubChem CID 2734320
IUPAC Name (4-tert-butylphenyl)boronic acid
SMILES CC(C)(C)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 191°C to 196°C
Quantity 1 g
Beilstein 5330959
Formula Weight 178.04
Percent Purity 97%
Chemical Name or Material 4-tert-Butylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.