Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylbenzenethiol 97.0+%, TCI America™
SDP

Catalog No. B074125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B074125G 25 g
B07415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B074125G Supplier TCI America Supplier No. B074125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2396-68-1
Molecular Formula C10H14S
Molecular Weight (g/mol) 166.282
MDL Number MFCD00022067
InChI Key GNXBFFHXJDZGEK-UHFFFAOYSA-N
Synonym 4-tert-butylthiophenol, p-tert-butylthiophenol, benzenethiol, p-tert-butyl, p-tert-butylbenzenethiol, p-t-butylthiophenol, p-tert-butylphenyl mercaptan, benzenethiol, 4-1,1-dimethylethyl, benzenethiol, p-tert-butylthio, 4-tert-butyl thiophenol, 4-t-butylthiophenol
PubChem CID 75454
IUPAC Name 4-tert-butylbenzenethiol
SMILES CC(C)(C)C1=CC=C(C=C1)S

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 166.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzenethiol
EINECSNumber (3)-1093
RTECSNumber DC1025000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.