Learn More
4-tert-Butylbenzoic acid, 99%
Description
4-tert-Butylbenzoic acid as a thermal stabilizer in PVC. It is used as a intermediate in chemical industry for producing esters of PTBBA, a chain stop agent in resins. It used as a potent yeast sirtuin (Sir2p) inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-tert-Butylbenzoic acid as a thermal stabilizer in PVC. It is used as a intermediate in chemical industry for producing esters of PTBBA, a chain stop agent in resins. It used as a potent yeast sirtuin (Sir2p) inhibitor.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 164°C to 168°C |
| Boiling Point | 280°C (decomposition) |
| Flash Point | 180°C (356°F) |
| Quantity | 5000 g |
| UN Number | UN2811 |
| Beilstein | 607545 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 178.23 |
| Percent Purity | ≥99% |
| Chemical Name or Material | 4-tert-Butylbenzoic acid |
Safety and Handling
| Hazard Category | H302-H311-H332-H360-H372-H500 |
| Hazard Statement | GHS H Statement H311-H360-H372-H373-H302 Toxic in contact with skin. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 202-696-3 |
| RTECSNumber | DG4708000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.