Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. B157625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B157625ML 25 mL
B15765ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B157625ML Supplier TCI America Supplier No. B157625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4210-32-6
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.232
MDL Number MFCD00075840
InChI Key IIZURLNRIMKEDL-UHFFFAOYSA-N
Synonym 4-tert-butyl benzonitrile, 4-t-butylbenzonitrile, benzonitrile, 4-1,1-dimethylethyl, 4-tert-butylcyanobenzene, 4-tert-butyl-benzonitrile, p-t-butylbenzonitrile, acmc-209jnk, p-tert-butylbenzonitrile, benzonitrile, p-tert-butyl, ksc219k6d
PubChem CID 77883
IUPAC Name 4-tert-butylbenzonitrile
SMILES CC(C)(C)C1=CC=C(C=C1)C#N

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 mL
UN Number 3276
Formula Weight 159.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.